Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6,7-Dibromobenzo(1,4)dioxan 97%

Supplier: AMBEED, INC
Warning

A342242-1G A342242-250MG A342242-5G
76773-162EA 15 USD InStock
76773-162 76773-164 76773-166
6,7-Dibromobenzo(1,4)dioxan 97%
6,7-Dibromobenzo(1,4)dioxan
Precaución: for Research Use Only


Formula: C₈H₆Br₂O₂
MDL Number: MFCD00463506
CAS Number: 25812-80-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty