Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Disopyramide 97%

Supplier: AMBEED, INC
Danger

A342795-10MG A342795-25MG A342795-50MG
76773-176EA 5 USD InStock
76773-176 76773-178 76773-180
Disopyramide 97%
Disopyramide
Precaución: for Research Use Only


Formula: C₂₁H₂₉N₃O
MW: 339.483 g/mol
MDL Number: MFCD00057366
CAS Number: 3737-09-5
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty