Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-amino-2-naphthoate 95%

Supplier: AMBEED, INC
Warning

A348769-100MG A348769-1G A348769-250MG A348769-5G
76773-262EA 10 USD InStock
76773-262 76773-264 76773-266 76773-268
Methyl-6-amino-2-naphthoate 95%
Methyl 6-amino-2-naphthoate
Precaución: for Research Use Only


Formula: C₁₂H₁₁NO₂
MW: 201.2200 g/mol
MDL Number: MFCD06797040
CAS Number: 5159-59-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty