Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-(trifluoromethyl)aniline 98%

Supplier: AMBEED, INC
Warning

A356340-10G A356340-25G A356340-500G A356340-100G A356340-5G
76773-308EA 6 USD InStock
76773-308 76773-310 76773-312 77626-688 77626-690
4-Chloro-2-(trifluoromethyl)aniline 98%
4-Chloro-2-(trifluoromethyl)aniline
Precaución: for Research Use Only


Formula: C₇H₅ClF₃N
MW: 195.57 g/mol
Boiling Pt: 213
Melting Pt: 9
MDL Number: MFCD00007841
CAS Number: 445-03-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty