Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Diphenylcarbamoyl chloride 98% (by GC)

Supplier: AMBEED, INC
Danger

A361611-25G A361611-5G A361611-100G
76773-348EA 14 USD InStock
76773-348 76773-350 77626-698
N,N-Diphenylcarbamoyl chloride 98% (by GC)
N,N-Diphenylcarbamoyl chloride
Precaución: for Research Use Only


Formula: C₁₃H₁₀ClNO
MDL Number: MFCD00000633
CAS Number: 83-01-2
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty