Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6,7-Dimethoxy-1-phenyl-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%

Supplier: AMBEED, INC
Warning

A365098-100MG A365098-1G A365098-250MG A365098-5G
76773-382EA 37 USD InStock
76773-382 76773-384 76773-386 76773-388
6,7-Dimethoxy-1-phenyl-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%
6,7-Dimethoxy-1-phenyl-1,2,3,4-tetrahydroisoquinoline hydrochloride
Precaución: for Research Use Only


Formula: C₁₇H₂₀ClNO₂
MW: 305.802 g/mol
MDL Number: MFCD00800512
CAS Number: 63768-20-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty