Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2-methylimidazo[1,2-a]pyridine 98%

Supplier: AMBEED, INC
Warning

A365348-1G A365348-250MG A365348-25G
76773-390EA 56 USD InStock
76773-390 76773-392 76773-394
6-Bromo-2-methylimidazo[1,2-a]pyridine 98%
6-Bromo-2-methylimidazo[1,2-a]pyridine
Precaución: for Research Use Only


Formula: C₈H₇BrN₂
MW: 211.07 g/mol
MDL Number: MFCD09800594
CAS Number: 4044-99-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty