Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Phenylsemicarbazide 98%

Supplier: AMBEED, INC
Warning

A368588-10G A368588-1G A368588-25G A368588-5G
76773-428EA 34 USD InStock
76773-428 76773-430 76773-432 76773-434
4-Phenylsemicarbazide 98%
4-Phenylsemicarbazide
Precaución: for Research Use Only


Formula: C₇H₉N₃O
MDL Number: MFCD00007590
CAS Number: 537-47-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty