Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2-pyridineboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A384272-10G A384272-1G A384272-250MG A384272-5G
76773-600EA 87 USD InStock
76773-600 76773-602 76773-604 77626-754
6-Bromo-2-pyridineboronic acid pinacol ester 98%
6-Bromo-2-pyridineboronic acid pinacol ester


Formula: C₁₁H₁₅BBrNO₂
MDL Number: MFCD06796177
CAS Number: 651358-83-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty