Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Phenyl-1-butanol 96%

Supplier: AMBEED, INC
Warning

A385985-10G A385985-1G A385985-25G A385985-5G
76773-628EA 157 USD InStock
76773-628 76773-630 76773-632 76773-634
1-Phenyl-1-butanol 96%
1-Phenyl-1-butanol
Precaución: for Research Use Only


Formula: C₁₀H₁₄O
MDL Number: MFCD00066173
CAS Number: 614-14-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty