Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(2-Fluorophenyl)-1,3,4-thiadiazol-2-amine 98%

Supplier: AMBEED, INC
Warning

A391894-100MG A391894-1G A391894-250MG A391894-5G
76773-652EA 25 USD InStock
76773-652 76773-654 76773-656 76773-658
5-(2-Fluorophenyl)-1,3,4-thiadiazol-2-amine 98%
5-(2-Fluorophenyl)-1,3,4-thiadiazol-2-amine
Precaución: for Research Use Only


Formula: C₈H₆FN₃S
MDL Number: MFCD00981035
CAS Number: 59565-51-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty