Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethylene diacrylate 90+% stabilised

Supplier: AMBEED, INC
Danger

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A392193-1G A392193-25G
76773-660EA 53 USD InStock
76773-660 77626-794
Ethylene diacrylate 90+% stabilised
Ethylene diacrylate
Precaución: for Research Use Only


Formula: C₈H₁₀O₄
MW: 170.164 g/mol
MDL Number: MFCD00008629
CAS Number: 2274-11-5
UN: 2810
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty