Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-tert-Butyl-2-methoxybenzeneboronic acid 95%

Supplier: AMBEED, INC
Warning

A412379-100MG A412379-1G A412379-250MG A412379-5G
76773-804EA 10 USD InStock
76773-804 76773-806 76773-808 76773-810
5-tert-Butyl-2-methoxybenzeneboronic acid 95%
5-tert-Butyl-2-methoxybenzeneboronic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₇BO₃
MW: 208.07 g/mol
MDL Number: MFCD06201034
CAS Number: 128733-85-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty