Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N,N',N'-Tetrakis(4-aminophenyl)-1,4-phenylenediamine 95%

Supplier: AMBEED, INC
Warning

A415671-100G A415671-5G A415671-1G
76773-840EA 392 USD InStock
76773-840 76773-842 77551-250
N,N,N',N'-Tetrakis(4-aminophenyl)-1,4-phenylenediamine 95%
N,N,N',N'-Tetrakis(4-aminophenyl)-1,4-phenylenediamine
Precaución: for Research Use Only


Formula: C₃₀H₂₈N₆
MDL Number: MFCD06657652
CAS Number: 3283-07-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty