Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-fluorophenetole 95%

Supplier: AMBEED, INC
Warning

A417074-1G A417074-250MG A417074-5G
76773-852EA 40 USD InStock
76773-852 76773-854 76773-856
5-Chloro-2-fluorophenetole 95%
5-Chloro-2-fluorophenetole
Precaución: for Research Use Only


Formula: C₈H₈ClFO
MW: 174.6 g/mol
MDL Number: MFCD00672980
CAS Number: 289039-34-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty