Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Morpholino-5-pyridineboronic acid 98%

Supplier: AMBEED, INC
Warning

A433867-10G A433867-1G A433867-250MG A433867-5G
76773-950EA 160 USD InStock
76773-950 76773-952 76773-954 76773-956
2-Morpholino-5-pyridineboronic acid 98%
2-Morpholino-5-pyridineboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₃BN₂O₃
MDL Number: MFCD03086116
CAS Number: 904326-93-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty