Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Monoethyl glutarate 98%

Supplier: AMBEED, INC
Warning

A452451-100G A452451-1G A452451-25G A452451-500G A452451-5G
76774-140EA 63 USD InStock
76774-140 76774-142 76774-144 76774-146 76774-148
Monoethyl glutarate 98%
Monoethyl glutarate
Precaución: for Research Use Only


Formula: C₇H₁₂O₄
MDL Number: MFCD00014379
CAS Number: 1070-62-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty