Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzo[f]quinoline 98% (by HPLC)

Supplier: AMBEED, INC
Danger

A479222-100MG A479222-1G
76774-342EA 69 USD InStock
76774-342 77626-954
Benzo[f]quinoline 98% (by HPLC)
Benzo[f]quinoline
Precaución: for Research Use Only


Formula: C₁₃H₉N
MDL Number: MFCD00022235
CAS Number: 85-02-9
UN: 2923
ADR: 8,6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty