Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromobenzo[b]thiophene-3-carbaldehyde 97%

Supplier: AMBEED, INC
Warning

A480591-100MG A480591-1G A480591-250MG A480591-5G
76774-358EA 8 USD InStock
76774-358 76774-360 76774-362 77626-956
5-Bromobenzo[b]thiophene-3-carbaldehyde 97%
5-Bromobenzo[b]thiophene-3-carbaldehyde
Precaución: for Research Use Only


Formula: C₉H₅BrOS
MW: 241.10 g/mol
MDL Number: MFCD00858578
CAS Number: 16296-72-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty