Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-tert-Butyl-2-cyanopyrrolidine-1-carboxylate 96%

Supplier: AMBEED, INC
Warning

A483590-100G A483590-10G A483590-1G A483590-250MG A483590-25G A483590-5G
76774-384EA 238 USD InStock
76774-384 76774-386 76774-388 76774-390 76774-392 76774-394
(S)-tert-Butyl-2-cyanopyrrolidine-1-carboxylate 96%
(S)-tert-Butyl-2-cyanopyrrolidine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₁₆N₂O₂
MW: 196.25 g/mol
MDL Number: MFCD01861224
CAS Number: 228244-04-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty