Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Hydroxy-2H-chromene-3-carboxaldehyde 97% (by HPLC)

Supplier: AMBEED, INC
Warning

A512800-100MG A512800-1G A512800-250MG A512800-5G
76774-570EA 24 USD InStock
76774-570 76774-572 76774-574 76774-576
6-Hydroxy-2H-chromene-3-carboxaldehyde 97% (by HPLC)
6-Hydroxy-2H-chromene-3-carboxaldehyde
Precaución: for Research Use Only


Formula: C₁₀H₈O₃
MDL Number: MFCD01076587
CAS Number: 134822-76-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty