Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,6-difluoroanisole 98%

Supplier: AMBEED, INC
Warning

A518131-1G A518131-250MG A518131-25G A518131-5G
76774-626EA 14 USD InStock
76774-626 76774-628 76774-630 77627-042
4-Bromo-2,6-difluoroanisole 98%
4-Bromo-2,6-difluoroanisole
Precaución: for Research Use Only


Formula: C₇H₅BrF₂O
MDL Number: MFCD01631351
CAS Number: 104197-14-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty