Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-2-methyl-2H-tetrazole 97%

Supplier: AMBEED, INC
Danger

A520707-1G A520707-5G
76774-648EA 19.1 USD InStock
76774-648 76774-650 76774-652
5-Amino-2-methyl-2H-tetrazole 97%
5-Amino-2-methyl-2H-tetrazole
Precaución: for Research Use Only


Formula: C₂H₅N₅
MDL Number: MFCD01819831
CAS Number: 6154-04-7
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty