Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium Dimethyl 5-sulfoisophthalate 98%

Supplier: AMBEED, INC
Warning

A524448-100G A524448-25G A524448-500G
76774-670EA 20 USD InStock
76774-670 76774-672 76774-674
Sodium Dimethyl 5-sulfoisophthalate 98%
Sodium Dimethyl 5-sulfoisophthalate
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₇S
MW: 296.22 g/mol
MDL Number: MFCD00007493
CAS Number: 3965-55-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty