Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4',5'-Dimethoxy-2'-hydroxyacetophenone 98%

Supplier: AMBEED, INC
Warning

A533356-1G A533356-250MG A533356-5G
76774-724EA 30 USD InStock
76774-724 76774-726 76774-728
4',5'-Dimethoxy-2'-hydroxyacetophenone 98%
4',5'-Dimethoxy-2'-hydroxyacetophenone
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₄
MW: 196.1999 g/mol
MDL Number: MFCD00017241
CAS Number: 20628-06-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty