Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Propylbenzylamine 97%

Supplier: AMBEED, INC
Warning

A544350-100MG A544350-1G A544350-250MG
76774-788EA 17 USD InStock
76774-788 76774-790 76774-792
4-Propylbenzylamine 97%
4-Propylbenzylamine
Precaución: for Research Use Only


Formula: C₁₀H₁₅N
MDL Number: MFCD03411009
CAS Number: 538342-98-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty