Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-(2-bromophenyl)-3-oxopropanoate 95%

Supplier: AMBEED, INC
Warning

A545445-100MG A545445-1G A545445-250MG A545445-5G
76774-798EA 9 USD InStock
76774-798 76774-800 76774-802 76774-804
Ethyl 3-(2-bromophenyl)-3-oxopropanoate 95%
Ethyl 3-(2-bromophenyl)-3-oxopropanoate
Precaución: for Research Use Only


Formula: C₁₁H₁₁BrO₃
MDL Number: MFCD03424813
CAS Number: 50671-05-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty