Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Isopropoxybenzoic acid 98%

Supplier: AMBEED, INC
Warning

A549363-100G A549363-25G A549363-5G
76774-818EA 131 USD InStock
76774-818 76774-820 76774-822
4-Isopropoxybenzoic acid 98%
4-Isopropoxybenzoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₃
MDL Number: MFCD00044318
CAS Number: 13205-46-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty