Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Fluoro-4-hydroxy-2-(trifluoromethyl)quinoline 98%

Supplier: AMBEED, INC
Warning

A550427-1G A550427-250MG A550427-25G A550427-5G
76774-824EA 23 USD InStock
76774-824 76774-826 76774-828 76774-830
8-Fluoro-4-hydroxy-2-(trifluoromethyl)quinoline 98%
8-Fluoro-4-hydroxy-2-(trifluoromethyl)quinoline
Precaución: for Research Use Only


Formula: C₁₀H₅F₄NO
MW: 231.15 g/mol
MDL Number: MFCD00276580
CAS Number: 31009-31-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty