Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(3-Pyridinyl)-1,3,4-oxadiazole-2-thiol 95%

Supplier: AMBEED, INC
Danger

A554602-1G A554602-250MG
76774-878EA 50 USD InStock
76774-878 76774-880
5-(3-Pyridinyl)-1,3,4-oxadiazole-2-thiol 95%
5-(3-Pyridinyl)-1,3,4-oxadiazole-2-thiol
Precaución: for Research Use Only


Formula: C₇H₅N₃OS
MDL Number: MFCD00139080
CAS Number: 3690-46-8
UN: 3259
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty