Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Nitro-1H-indole-2-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A556066-100MG A556066-1G A556066-250MG
76774-884EA 23 USD InStock
76774-884 76774-886 76774-888
6-Nitro-1H-indole-2-carboxylic acid 95%
6-Nitro-1H-indole-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₉H₆N₂O₄
MDL Number: MFCD02664466
CAS Number: 10242-00-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty