Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethoxy)benzaldehyde 95%

Supplier: AMBEED, INC
Warning

A565545-100G A565545-10G A565545-1G A565545-25G A565545-500G A565545-5G
76774-976EA 57 USD InStock
76774-976 76774-978 76774-980 76774-982 76774-984 76774-986
4-(Trifluoromethoxy)benzaldehyde 95%
4-(Trifluoromethoxy)benzaldehyde
Precaución: for Research Use Only


Formula: C₈H₅F₃O₂
MDL Number: MFCD00041530
CAS Number: 659-28-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty