Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-4-methoxybenzonitrile 98%

Supplier: AMBEED, INC
Warning

A577603-100G A577603-10G A577603-1G A577603-25G A577603-5G
76775-024EA 155 USD InStock
76775-024 76775-026 76775-028 76775-030 76775-032
3-Bromo-4-methoxybenzonitrile 98%
3-Bromo-4-methoxybenzonitrile
Precaución: for Research Use Only


Formula: C₈H₆BrNO
MDL Number: MFCD00143097
CAS Number: 117572-79-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty