Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3-(biphenyl-4-yl)propanoic acid 95%

Supplier: AMBEED, INC
Warning

A589247-1G A589247-250MG A589247-5G
76775-104EA 87 USD InStock
76775-104 76775-106 76775-108
2-Amino-3-(biphenyl-4-yl)propanoic acid 95%
2-Amino-3-(biphenyl-4-yl)propanoic acid
Precaución: for Research Use Only


Formula: C₁₅H₁₅NO₂
MW: 241.285 g/mol
MDL Number: MFCD03425234
CAS Number: 76985-08-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty