Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Fluorochromone 98%

Supplier: AMBEED, INC
Warning

A589563-1G A589563-250MG A589563-5G
76775-116EA 18 USD InStock
76775-116 76775-118 76775-120
6-Fluorochromone 98%
6-Fluorochromone
Precaución: for Research Use Only


Formula: C₉H₅FO₂
MDL Number: MFCD03094002
CAS Number: 105300-38-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty