Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Pentafluorophenyl)acetonitrile 98%

Supplier: AMBEED, INC
Danger

A611050-1G A611050-250MG A611050-5G
76775-296EA 27 USD InStock
76775-296 76775-298 76775-300
(Pentafluorophenyl)acetonitrile 98%
(Pentafluorophenyl)acetonitrile
Precaución: for Research Use Only


Formula: (C₆F₅)CH₂CN
MDL Number: MFCD00013814
CAS Number: 653-30-5
UN: 3276
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty