Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Neochlorogenic acid 98%

Supplier: AMBEED, INC
Warning

A612121-10MG A612121-1MG A612121-5MG
76775-302EA 100 USD InStock
76775-302 76775-304 76775-306
Neochlorogenic acid 98%
Neochlorogenic acid
Precaución: for Research Use Only


Formula: C₁₆H₁₈O₉
MDL Number: MFCD10566639
CAS Number: 906-33-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty