Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-methyl-5-isoxazolamine 98%

Supplier: AMBEED, INC
Warning

A616046-1G A616046-250MG A616046-25G A616046-5G
76775-338EA 5.31 USD InStock
76775-338 76775-340 76775-342 76775-344 76775-346
4-Chloro-3-methyl-5-isoxazolamine 98%
4-Chloro-3-methyl-5-isoxazolamine
Precaución: for Research Use Only


Formula: C₄H₅ClN₂O
MDL Number: MFCD08443950
CAS Number: 166964-09-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty