Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-Amino-3-(4-hydroxyphenyl)-2-methylpropanoic acid 95%

Supplier: AMBEED, INC
Warning

A624425-100MG A624425-10MG A624425-1G A624425-1MG A624425-250MG A624425-25MG A624425-50MG A624425-5MG A624425-5G
76775-402EA 31 USD InStock
76775-402 76775-404 76775-406 76775-408 76775-410 76775-412 76775-414 76775-416 77627-256
(S)-2-Amino-3-(4-hydroxyphenyl)-2-methylpropanoic acid 95%
(S)-2-Amino-3-(4-hydroxyphenyl)-2-methylpropanoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₃NO₃
MDL Number: MFCD00064201
CAS Number: 672-87-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty