Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-tert-Butylcyclohexanone 98%

Supplier: AMBEED, INC
Warning

A625676-1000G A625676-100G A625676-1G A625676-25G A625676-500G A625676-5G
76775-432EA 233 USD InStock
76775-432 76775-434 76775-436 76775-438 76775-440 76775-442
4-tert-Butylcyclohexanone 98%
4-tert-Butylcyclohexanone
Precaución: for Research Use Only


Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 225
MDL Number: MFCD00001642
CAS Number: 98-53-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty