Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Difluorobenzyl alcohol 96%

Supplier: AMBEED, INC
Warning

A627298-10G A627298-1G A627298-25G A627298-5G
76775-446EA 17 USD InStock
76775-446 76775-448 76775-450 76775-452
2,4-Difluorobenzyl alcohol 96%
2,4-Difluorobenzyl alcohol
Precaución: for Research Use Only


Formula: C₇H₆F₂O
MDL Number: MFCD00009983
CAS Number: 56456-47-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty