Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-methoxypropionate 95%

Supplier: AMBEED, INC
Danger

A638490-25G A638490-5G
76775-488EA 10 USD InStock
76775-488 76775-490
Methyl-3-methoxypropionate 95%
Methyl 3-methoxypropionate
Precaución: for Research Use Only


Formula: C₅H₁₀O₃
MDL Number: MFCD00059085
CAS Number: 3852-09-3
UN: 3272
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty