Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Morpholinoacetophenone 98%

Supplier: AMBEED, INC
Warning

A640860-25G A640860-5G
76775-500EA 58 USD InStock
76775-500 76775-502
4'-Morpholinoacetophenone 98%
4'-Morpholinoacetophenone
Precaución: for Research Use Only


Formula: C₁₂H₁₅NO₂
MDL Number: MFCD00006168
CAS Number: 39910-98-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty