Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxyphenacyl thiocyanate 95%

Supplier: AMBEED, INC
Warning

A658249-1G A658249-250MG A658249-5G
76775-610EA 54 USD InStock
76775-610 76775-612 76775-614
4-Methoxyphenacyl thiocyanate 95%
4-Methoxyphenacyl thiocyanate
Precaución: for Research Use Only


Formula: C₁₀H₉NO₂S
MDL Number: MFCD01332833
CAS Number: 6097-26-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty