Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-1-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile 95%

Supplier: AMBEED, INC
Danger

A663433-1G A663433-250MG A663433-5G
76775-648EA 27 USD InStock
76775-648 76775-650 76775-652
5-Amino-1-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile 95%
5-Amino-1-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile
Precaución: for Research Use Only


Formula: C₁₀H₇ClN₄
MDL Number: MFCD00052031
CAS Number: 51516-67-7
UN: 3259
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty