Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N'-Diisopropyl-O-(4-nitrobenzyl)isourea 96%

Supplier: AMBEED, INC
Warning

A665945-1G A665945-25G A665945-5G
76775-654EA 21 USD InStock
76775-654 76775-656 76775-658
N,N'-Diisopropyl-O-(4-nitrobenzyl)isourea 96%
N,N'-Diisopropyl-O-(4-nitrobenzyl)isourea
Precaución: for Research Use Only


Formula: C₁₄H₂₁N₃O₃
MW: 279.34 g/mol
MDL Number: MFCD00042046
CAS Number: 2978-11-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty