Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-mercaptopropionate 99%

Supplier: AMBEED, INC
Danger

A685507-25G A685507-5G
76775-770EA 29 USD InStock
76775-770 76775-772
Ethyl-3-mercaptopropionate 99%
Ethyl-3-mercaptopropionate
Precaución: for Research Use Only


Formula: C₅H₁₀O₂S
MDL Number: MFCD00004896
CAS Number: 5466-06-8
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty