Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-o-cresol 98%

Supplier: AMBEED, INC
Warning

A688326-100G A688326-10G A688326-1G A688326-25G A688326-5G
76775-800EA 83 USD InStock
76775-800 76775-802 76775-804 76775-806 76775-808
5-Fluoro-o-cresol 98%
5-Fluoro-o-cresol
Precaución: for Research Use Only


Formula: C₇H₇FO
MW: 126.13 g/mol
MDL Number: MFCD00673009
CAS Number: 452-85-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty