Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid 98%

Supplier: AMBEED, INC
Warning

A695706-1G A695706-250MG A695706-25G A695706-5G
76775-870EA 47 USD InStock
76775-870 76775-872 76775-874 76775-876
(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid 98%
(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₅
MW: 212.2
MDL Number: MFCD00016834
CAS Number: 4385-56-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty