Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethyl)cinnamic acid 98%

Supplier: AMBEED, INC
Warning

A698959-100G A698959-10G A698959-1G A698959-5G
76775-900EA 282 USD InStock
76775-900 76775-902 76775-904 76775-906
2-(Trifluoromethyl)cinnamic acid 98%
2-(Trifluoromethyl)cinnamic acid
Precaución: for Research Use Only


Formula: C₁₀H₇F₃O₂
MW: 216.16
Boiling Pt: 292
Melting Pt: 199
MDL Number: MFCD00004381
CAS Number: 2062-25-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty